(Z)-N'-Hydroxy-2-[4-oxo-2-(thiophen-2-yl)-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]ethenimidamide

Supplier: Apollo Scientific

OR311005-1G OR311005-500MG
APOSOR311005-1GEA 539 EUR
APOSOR311005-1G APOSOR311005-500MG
(Z)-N'-Hydroxy-2-[4-oxo-2-(thiophen-2-yl)-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]ethenimidamide
(Z)-N'-Hydroxy-2-[4-oxo-2-(thiophen-2-yl)-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]ethenimidamide
Formula: C₁₂H₁₁N₅O₂S
MW: 289,313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17780264
CAS Number: 1255791-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR