3-Phenyl-1,4-diazaspiro[4.5]dec-3-ene-2-thione

Supplier: Apollo Scientific

OR311011-1G OR311011-500MG
APOSOR311011-1GEA 539 EUR
APOSOR311011-1G APOSOR311011-500MG
3-Phenyl-1,4-diazaspiro[4.5]dec-3-ene-2-thione
3-Phenyl-1,4-diazaspiro[4.5]dec-3-ene-2-thione
Formula: C₁₄H₁₆N₂S
MW: 244,355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02311198
CAS Number: 4602-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR