N-Phenyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide

Supplier: Apollo Scientific

OR311013-1G OR311013-500MG
APOSOR311013-1GEA 539 EUR
APOSOR311013-1G APOSOR311013-500MG
N-Phenyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
N-Phenyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
Formula: C₁₆H₁₃N₃OS
MW: 295,359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14703430
CAS Number: 1193387-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR