2-(4-Methoxyphenyl)-4-phenyl-1H-imidazole-5-thiol
Supplier: Apollo Scientific
OR311024-1G
OR311024-500MG
APOSOR311024-1GEA
539
EUR
APOSOR311024-1G
APOSOR311024-500MG
2-(4-Methoxyphenyl)-4-phenyl-1H-imidazole-5-thiol
2-(4-Methoxyphenyl)-4-phenyl-1H-imidazole-5-thiol
Formula:
C₁₆H₁₄N₂OS MW: 282,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22192621 CAS Number: 1325304-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...