3-Butanoyl-8-methoxy-2H-chromen-2-one

Supplier: Apollo Scientific

OR311036-1G OR311036-500MG
APOSOR311036-1GEA 539 EUR
APOSOR311036-1G APOSOR311036-500MG
3-Butanoyl-8-methoxy-2H-chromen-2-one
3-Butanoyl-8-methoxy-2H-chromen-2-one
Formula: C₁₄H₁₄O₄
MW: 246,259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18203429
CAS Number: 100886-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR