Ethyl 6-bromo-1-benzofuran-2-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 6-bromobenzofuran-2-carboxylate

OR311054-1G OR311054-250MG
APOSOR311054-1GEA 287 EUR
APOSOR311054-1G APOSOR311054-250MG
Ethyl 6-bromo-1-benzofuran-2-carboxylate
Ethyl 6-bromo-1-benzofuran-2-carboxylate
Formula: C₁₁H₉BrO₃
MW: 269,091 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD16038162
CAS Number: 907945-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR