7-(Phenylcarbamoyl)heptanoic acid

Supplier: Apollo Scientific
Warning

OR311141-1G OR311141-500MG
APOSOR311141-1GEA 671 EUR
APOSOR311141-1G APOSOR311141-500MG
7-(Phenylcarbamoyl)heptanoic acid
7-(Phenylcarbamoyl)heptanoic acid
Formula: C₁₄H₁₉NO₃
MW: 249,306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07777004
CAS Number: 149648-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR