2-Ethyl-2-methylbutanoic acid

Supplier: Apollo Scientific
Warning

OR311177-1G OR311177-250MG
APOSOR311177-1GEA 655 EUR
APOSOR311177-1G APOSOR311177-250MG
2-Ethyl-2-methylbutanoic acid
2-Ethyl-2-methylbutanoic acid
Formula: C₇H₁₄O₂
MW: 130,185 g/mol
Boiling Pt: 200 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00055649
CAS Number: 19889-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR