Bicyclo[1.1.1]pentane-1-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR312074-250MG
OR312074-100MG
APOSOR312074-250MGEA
129
EUR
APOSOR312074-250MG
APOSOR312074-100MG
Bicyclo[1.1.1]pentane-1-carboxylic acid
Bicyclo[1.1.1]pentane-1-carboxylic acid
Formula:
C₆H₈O₂ Boiling Pt: 125…127 °C (45 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD19228106 CAS Number: 22287-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...