Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR312113-1G
OR312113-250MG
APOSOR312113-1GEA
967
EUR
APOSOR312113-1G
APOSOR312113-250MG
Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride
Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride
Formula:
C₅H₁₀N₂·2ClH MW: 171,068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19211809 CAS Number: 147927-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...