4-Formylbenzenesulphonyl chloride 95%

Supplier: Apollo Scientific
Danger

OR313065-1G OR313065-5G
APOSOR313065-1GEA 161 EUR
APOSOR313065-1G APOSOR313065-5G
4-Formylbenzenesulphonyl chloride 95%
4-Formylbenzenesulphonyl chloride
Formula: C₇H₅ClO₃S
Storage Temperature: Ambient
MDL Number: MFCD01861215
CAS Number: 85822-16-8
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR