(S)-3-Methyl-pyrrolidine hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: (S)-3-Methylpyrrolidine hydrochloride

OR313067-1G OR313067-250MG
APOSOR313067-1GEA 222 EUR
APOSOR313067-1G APOSOR313067-250MG
(S)-3-Methyl-pyrrolidine hydrochloride
(S)-3-Methyl-pyrrolidine hydrochloride
Formula: C₅H₁₁N·ClH
MW: 121,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796632
CAS Number: 186597-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR