Cyclobutanecarboxaldehyde

Supplier: Apollo Scientific
Warning

OR313071-10G OR313071-1G OR313071-5G
APOSOR313071-10GEA 0 EUR
APOSOR313071-10G APOSOR313071-1G APOSOR313071-5G
Cyclobutanecarboxaldehyde
Cyclobutanecarboxaldehyde
Formula: C₅H₈O
Melting Pt: 115…117 °C
Density: 0,9355 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03789631
CAS Number: 2987-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR