Isothiazole-5-carboxylic acid

Supplier: Apollo Scientific

OR313078-1G OR313078-250MG
APOSOR313078-1GEA 192 EUR
APOSOR313078-1G APOSOR313078-250MG
Isothiazole-5-carboxylic acid
Isothiazole-5-carboxylic acid
Formula: C₄H₃NO₂S
MW: 129,137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369964
CAS Number: 10271-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR