4-Pyridineacetic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR313081-5G
APOSOR313081-5GEA
162
EUR
APOSOR313081-5G
4-Pyridineacetic acid 95%
4-Pyridineacetic acid
Formula:
C₇H₇NO₂ MW: 137,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701446 CAS Number: 28356-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...