7-Chloro-1-indanone

Supplier: Apollo Scientific
Warning

Synonyms: 7-Chlorindan-1-on

OR315162-10G OR315162-1G OR315162-5G
APOSOR315162-10GEA 364 EUR
APOSOR315162-10G APOSOR315162-1G APOSOR315162-5G
7-Chloro-1-indanone
7-Chloro-1-indanone
Formula: C₉H₇ClO
MW: 166,604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796576
CAS Number: 34911-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR