Phenylcyanate

Supplier: Apollo Scientific

Synonyms: Phenyl Cyanate

OR315259-1G OR315259-5G
APOSOR315259-1GEA 43.5 EUR
APOSOR315259-1G APOSOR315259-5G
Phenylcyanate
Phenylcyanate
Formula: C₇H₅NO
MW: 119,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973644
CAS Number: 1122-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR