3-Cyclopropylthiophene-4-carboxaldehyde

Supplier: Apollo Scientific

OR315856-1G OR315856-250MG
APOSOR315856-1GEA 655 EUR
APOSOR315856-1G APOSOR315856-250MG
3-Cyclopropylthiophene-4-carboxaldehyde
3-Cyclopropylthiophene-4-carboxaldehyde
Formula: C₈H₈OS
MW: 152,214 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22032411
CAS Number: 1369114-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR