3-Cyclopropylthiophene-4-carbonitrile 95%
Supplier: Apollo Scientific
OR315859-1G
OR315859-250MG
APOSOR315859-1GEA
574
EUR
APOSOR315859-1G
APOSOR315859-250MG
3-Cyclopropylthiophene-4-carbonitrile 95%
3-Cyclopropylthiophene-4-carbonitrile
Formula:
C₈H₇NS MW: 149,213 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30074923 CAS Number: 1482683-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...