6-Cyclopropylpyridine-2-carbaldehyde 95%

Supplier: Apollo Scientific

OR315866-1G OR315866-250MG
APOSOR315866-1GEA 633 EUR
APOSOR315866-1G APOSOR315866-250MG
6-Cyclopropylpyridine-2-carbaldehyde 95%
6-Cyclopropylpyridine-2-carbaldehyde
Formula: C₉H₉NO
MW: 147,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255567
CAS Number: 208111-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR