4-(Oxetan-3-yl)piperidine hemioxalate
Supplier: Apollo Scientific
|
Warning
|
OR317045-1G
OR317045-250MG
OR317045-500MG
APOSOR317045-1GEA
1480
EUR
APOSOR317045-1G
APOSOR317045-250MG
APOSOR317045-500MG
4-(Oxetan-3-yl)piperidine hemioxalate
4-(Oxetan-3-yl)piperidine hemioxalate
Formula:
2C₈H₁₅NO·C₂H₂O₄ MW: 372,456 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23105786 CAS Number: 1523606-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...