1-Amino-5-bromonaphthalene 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromonaphthalen-1-amine, 5-Bromo-1-naphthylamine

OR3180-25G OR3180-5G
APOSOR3180-25GEA 130 EUR
APOSOR3180-25G APOSOR3180-5G
1-Amino-5-bromonaphthalene 95%
1-Amino-5-bromonaphthalene
Formula: C₁₀H₈BrN
MW: 222.081 g/mol
Boiling Pt: 180…185 °C (7 mm Hg)
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD07369883
CAS Number: 4766-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR