tert-Butyl 3-(benzylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%

Supplier: Apollo Scientific

OR318034-1G OR318034-5G
APOSOR318034-1GEA 441 EUR
APOSOR318034-1G APOSOR318034-5G
tert-Butyl 3-(benzylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%
tert-Butyl 3-(benzylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate
Formula: C₁₉H₂₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22690719
CAS Number: 944123-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR