2-Methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%

Supplier: Apollo Scientific

OR32080-1G OR32080-5G
APOSOR32080-1GEA 328 EUR
APOSOR32080-1G APOSOR32080-5G
2-Methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%
2-Methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione
Formula: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00770324
CAS Number: 4494-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR