Methyl (2E)-3-(3-bromophenyl)prop-2-enoate 95%

Supplier: Apollo Scientific

Synonyms: (E)-Methyl 3-(3-bromophenyl)acrylate

OR32116-1G OR32116-5G
APOSOR32116-1GEA 154 EUR
APOSOR32116-1G APOSOR32116-5G
Methyl (2E)-3-(3-bromophenyl)prop-2-enoate 95%
Methyl (2E)-3-(3-bromophenyl)prop-2-enoate
Formula: C₁₀H₉BrO₂
MW: 241,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00296965
CAS Number: 79432-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR