3-(Piperidin-4-yl)pyridine dihydrochloride

Supplier: Apollo Scientific

OR321399-1G OR321399-250MG
APOSOR321399-1GEA 655 EUR
APOSOR321399-1G APOSOR321399-250MG
3-(Piperidin-4-yl)pyridine dihydrochloride
3-(Piperidin-4-yl)pyridine dihydrochloride
Formula: C₁₀H₁₄N₂·2ClH
Storage Temperature: Refrigerator
MDL Number: MFCD26406202
CAS Number: 301222-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR