4-(Piperidin-4-yl)isoquinoline dihydrochloride

Supplier: Apollo Scientific

OR321413-1G OR321413-250MG
APOSOR321413-1GEA 647 EUR
APOSOR321413-1G APOSOR321413-250MG
4-(Piperidin-4-yl)isoquinoline dihydrochloride
4-(Piperidin-4-yl)isoquinoline dihydrochloride
Formula: C₁₄H₁₆N₂·2ClH
MW: 285,212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31629927
CAS Number: 2244087-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR