(S)-2-(Pyrrolidin-2-yl)pyridine dihydrochloride
Supplier: Apollo Scientific
OR321537-1G
OR321537-250MG
APOSOR321537-1GEA
563
EUR
APOSOR321537-1G
APOSOR321537-250MG
(S)-2-(Pyrrolidin-2-yl)pyridine dihydrochloride
(S)-2-(Pyrrolidin-2-yl)pyridine dihydrochloride
Formula:
C₉H₁₂N₂·2ClH MW: 221,127 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30735892 CAS Number: 2097073-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...