Ethyl 4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 95%
Supplier: Apollo Scientific
OR32219-1G
OR32219-5G
APOSOR32219-1GEA
328
EUR
APOSOR32219-1G
APOSOR32219-5G
Ethyl 4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 95%
Ethyl 4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate
Formula:
C₁₁H₁₀N₂O₃ MW: 218,209 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02102304 CAS Number: 32092-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...