5-Bromobenzo[d]oxazol-2-amine

Supplier: Apollo Scientific
Warning

OR322634-25G
APOSOR322634-25GEA 185 EUR
APOSOR322634-25G
5-Bromobenzo[d]oxazol-2-amine
5-Bromobenzo[d]oxazol-2-amine
Formula: C₇H₅BrN₂O
MW: 213,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01664213
CAS Number: 64037-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR