4-(Chloromethyl)benzaldehyde
Supplier: Apollo Scientific
|
Danger
|
OR322751-1G
OR322751-5G
OR322751-25G
APOSOR322751-1GEA
23.4
EUR
APOSOR322751-1G
APOSOR322751-5G
APOSOR322751-25G
4-(Chloromethyl)benzaldehyde
4-(Chloromethyl)benzaldehyde
Formula:
C₈H₇ClO MW: 154,594 g/mol Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191396 CAS Number: 73291-09-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...