2-Iodo-4-nitrobenzoic acid 95%

Supplier: Apollo Scientific

OR322775-1G OR322775-250MG OR322775-5G
APOSOR322775-1GEA 30.6 EUR
APOSOR322775-1G APOSOR322775-250MG APOSOR322775-5G
2-Iodo-4-nitrobenzoic acid 95%
2-Iodo-4-nitrobenzoic acid
Formula: C₇H₄INO₄
MW: 293,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00956495
CAS Number: 89459-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR