Methyl 3-amino-3-(naphthalen-1-yl)propanoate, tech

Supplier: Apollo Scientific

OR32340-1G
APOSOR32340-1GEA 1570 EUR
APOSOR32340-1G
Methyl 3-amino-3-(naphthalen-1-yl)propanoate, tech
Methyl 3-amino-3-(naphthalen-1-yl)propanoate
Formula: C₁₄H₁₅NO₂
MW: 229,274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03011782
CAS Number: 188812-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR