N-[2-(3,4-Dimethoxyphenyl)ethyl]formamide 95%
Supplier: Apollo Scientific
OR32477-1G
OR32477-5G
APOSOR32477-1GEA
328
EUR
APOSOR32477-1G
APOSOR32477-5G
N-[2-(3,4-Dimethoxyphenyl)ethyl]formamide 95%
N-[2-(3,4-Dimethoxyphenyl)ethyl]formamide
Formula:
C₁₁H₁₅NO₃ MW: 209,242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03012208 CAS Number: 14301-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...