(2E,4E)-5-(2H-1,3-Benzodioxol-5-yl)penta-2,4-dienoic acid 95%

Supplier: Apollo Scientific

Synonyms: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)penta-2,4-dienoic acid

OR32511-1G OR32511-5G
APOSOR32511-1GEA 168 EUR
APOSOR32511-1G APOSOR32511-5G
(2E,4E)-5-(2H-1,3-Benzodioxol-5-yl)penta-2,4-dienoic acid 95%
(2E,4E)-5-(2H-1,3-Benzodioxol-5-yl)penta-2,4-dienoic acid
Formula: C₁₂H₁₀O₄
MW: 218,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047282
CAS Number: 136-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR