2-Chloro-3-(1H-pyrrol-1-yl)pyridine 95%
Supplier: Apollo Scientific
OR33430-1G
OR33430-5G
APOSOR33430-1GEA
109
EUR
APOSOR33430-1G
APOSOR33430-5G
2-Chloro-3-(1H-pyrrol-1-yl)pyridine 95%
2-Chloro-3-(1H-pyrrol-1-yl)pyridine
Formula:
C₉H₇ClN₂ MW: 178,618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084976 CAS Number: 70291-26-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...