6-Bromo-2,3-diaminopyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromopyridine-2,3-diamine
OR345244-1G
APOSOR345244-1GEA
65.4
EUR
APOSOR345244-1G
6-Bromo-2,3-diaminopyridine
6-Bromo-2,3-diaminopyridine
Formula:
C₅H₆BrN₃ MW: 188,025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10697695 CAS Number: 129012-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...