2-(Morpholin-4-yl)acetamidoxime

Supplier: Apollo Scientific

OR346064-1G OR346064-5G
APOSOR346064-1GEA 0 EUR
APOSOR346064-1G APOSOR346064-5G
2-(Morpholin-4-yl)acetamidoxime
2-(Morpholin-4-yl)acetamidoxime
Formula: C₆H₁₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09948347
CAS Number: 5815-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR