3-(Morpholin-4-yl)propionamidoxime
Supplier: Apollo Scientific
OR346065-1G
OR346065-5G
APOSOR346065-1GEA
0
EUR
APOSOR346065-1G
APOSOR346065-5G
3-(Morpholin-4-yl)propionamidoxime
3-(Morpholin-4-yl)propionamidoxime
Formula:
C₇H₁₅N₃O₂ MW: 173,213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03033671 CAS Number: 350682-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...