3-Methyl-isothiazole-5-carbonitrile
Supplier: Apollo Scientific
OR346141-1G
OR346141-500MG
APOSOR346141-1GEA
1050
EUR
APOSOR346141-1G
APOSOR346141-500MG
3-Methyl-isothiazole-5-carbonitrile
3-Methyl-isothiazole-5-carbonitrile
Formula:
C₅H₄N₂S MW: 124,164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110312 CAS Number: 57352-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...