3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester

Supplier: Apollo Scientific

Synonyms: Ethyl 3-cyclopropyl-1,2,4-oxadiazole-5-carboxylate

OR346203-1G OR346203-500MG
APOSOR346203-1GEA 0 EUR
APOSOR346203-1G APOSOR346203-500MG
3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester
3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester
Formula: C₈H₁₀N₂O₃
MW: 182,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701787
CAS Number: 1018125-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR