5-Hydroxy-1H-pyrazole-3-carboxylic acid

Supplier: Apollo Scientific
Warning

OR346229-1G OR346229-5G
APOSOR346229-1GEA 127 EUR
APOSOR346229-1G APOSOR346229-5G
5-Hydroxy-1H-pyrazole-3-carboxylic acid
5-Hydroxy-1H-pyrazole-3-carboxylic acid
Formula: C₄H₄N₂O₃
MW: 128,086 g/mol
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD12195917
CAS Number: 89603-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR