1-(2-Bromo-ethyl)-piperidine hydrobromide

Supplier: Apollo Scientific

Synonyms: 1-(2-Bromoethyl)piperidine hydrobromide

OR346238-1G OR346238-5G
APOSOR346238-1GEA 155 EUR
APOSOR346238-1G APOSOR346238-5G
1-(2-Bromo-ethyl)-piperidine hydrobromide
1-(2-Bromo-ethyl)-piperidine hydrobromide
Formula: C₇H₁₄BrN·BrH
MW: 273,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463352
CAS Number: 89796-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR