5-Ethyl-2-methyl-nicotinic acid
Supplier: Apollo Scientific
|
Warning
|
OR346243-1G
OR346243-5G
APOSOR346243-1GEA
0
EUR
APOSOR346243-1G
APOSOR346243-5G
5-Ethyl-2-methyl-nicotinic acid
5-Ethyl-2-methyl-nicotinic acid
Formula:
C₉H₁₁NO₂ MW: 165,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18259664 CAS Number: 22701-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...