4-Nitro-piperidine-1-carboxylic acid tert-butyl ester

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 4-nitropiperidine-1-carboxylate

OR346372-1G OR346372-5G
APOSOR346372-1GEA 69.6 EUR
APOSOR346372-1G APOSOR346372-5G
4-Nitro-piperidine-1-carboxylic acid tert-butyl ester
4-Nitro-piperidine-1-carboxylic acid tert-butyl ester
Formula: C₁₀H₁₈N₂O₄
MW: 230,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618559
CAS Number: 1228630-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR