6-Methyl-1H-indole-2-carboxylic acid 95+%

Supplier: Apollo Scientific

Synonyms: eIF4A3-IN-8

OR346380-1G OR346380-5G
APOSOR346380-1GEA 161 EUR
APOSOR346380-1G APOSOR346380-5G
6-Methyl-1H-indole-2-carboxylic acid 95+%
6-Methyl-1H-indole-2-carboxylic acid
Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664477
CAS Number: 18474-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR