3,5-Dibromo-dihydro-pyran-2,6-dione 95+%

Supplier: Apollo Scientific

OR346394-1G OR346394-500MG
APOSOR346394-1GEA 317 EUR
APOSOR346394-1G APOSOR346394-500MG
3,5-Dibromo-dihydro-pyran-2,6-dione 95+%
3,5-Dibromo-dihydro-pyran-2,6-dione
Formula: C₅H₄Br₂O₃
MW: 271,891 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053009
CAS Number: 121049-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR