1-(3-Chlorophenyl)-1H-pyrazole 95+%

Supplier: Apollo Scientific

OR346395-1G OR346395-5G
APOSOR346395-1GEA 154 EUR
APOSOR346395-1G APOSOR346395-5G
1-(3-Chlorophenyl)-1H-pyrazole 95+%
1-(3-Chlorophenyl)-1H-pyrazole
Formula: C₉H₇ClN₂
MW: 178,618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845709
CAS Number: 57211-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR