(Z)-3-Amino-3-(3-bromophenyl)acrylonitrile 95+%

Supplier: Apollo Scientific

OR346416-1G OR346416-5G
APOSOR346416-1GEA 201 EUR
APOSOR346416-1G APOSOR346416-5G
(Z)-3-Amino-3-(3-bromophenyl)acrylonitrile 95+%
(Z)-3-Amino-3-(3-bromophenyl)acrylonitrile
Formula: C₉H₇BrN₂
MW: 223,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12131143
CAS Number: 96017-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR