(Z)-3-Amino-3-(4-bromophenyl)acrylonitrile 95+%

Supplier: Apollo Scientific

OR346417-1G OR346417-5G
APOSOR346417-1GEA 201 EUR
APOSOR346417-1G APOSOR346417-5G
(Z)-3-Amino-3-(4-bromophenyl)acrylonitrile 95+%
(Z)-3-Amino-3-(4-bromophenyl)acrylonitrile
Formula: C₉H₇BrN₂
MW: 223,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026370
CAS Number: 71682-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR